Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBOX5 Monoclonal Antibody (OTI5G9), TrueMAB™, OriGene
SDP

Catalog No. 502541204
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1204 30 μL
50-167-3947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1204 Supplier OriGene Supplier No. TA502952S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBOX5 Monoclonal antibody specifically detects UBOX5 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen UBOX5
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI5G9
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Lgr5
Gene Accession No. O75473
Gene Alias 1500010O06Rik; C330018L13Rik; FAST kinase domain-containing protein 5; RING finger protein 37; FAST kinase domains 5; FASTKD5; hUIP5; KIAA0860; RGD1305440; RING finger protein 37; RING-type E3 ubiquitin transferase RNF37; Rnf37; U box domain containing 5; UBCE7IP5; ubcM4-interacting protein 5; Ube7ip5; ubiquitin conjugating enzyme 7 interacting protein 5; ubiquitin-conjugating enzyme 7-interacting protein 5; U-box domain containing 5; U-box domain-containing protein 5; Ubox5; UIP5; Unknown (protein for MGC:138103)
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 250-550 of human LGR5 produced in 293T Cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8549
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.