Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBOX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555995 Supplier Invitrogen™ Supplier No. PA555995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTLVGKVLLK NQSQVVFSHR GFKARPPFGA MEATLPSPAV VAQELWNKGA LSLSHVAHLR ICITHVTGGG IPCIKRLEVW GQPAKTCSQE VIDSILLVTS ENLPQDVALQ APALPMESDC DP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.

UBOX5 encodes a U-box domain containing protein. The encoded protein interacts with E2 enzymes and may play a role in the ubiquitination pathway. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBOX5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ubox5
Gene Accession No. O94941
Gene Alias 1500010O06Rik; C330018L13Rik; FAST kinase domain-containing protein 5; RING finger protein 37; FAST kinase domains 5; FASTKD5; hUIP5; KIAA0860; RGD1305440; RING finger protein 37; RING-type E3 ubiquitin transferase RNF37; Rnf37; U box domain containing 5; UBCE7IP5; ubcM4-interacting protein 5; Ube7ip5; ubiquitin conjugating enzyme 7 interacting protein 5; ubiquitin-conjugating enzyme 7-interacting protein 5; U-box domain containing 5; U-box domain-containing protein 5; Ubox5; UIP5; Unknown (protein for MGC:138103)
Gene Symbols Ubox5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBCE7IP5. Recombinant protein control fragment (Product #RP-102078).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.