Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBOX5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988853
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8853 Supplier Bioss Supplier No. BS9145R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBOX5 encodes a U-box domain containing protein. The encoded protein interacts with E2 enzymes and may play a role in the ubiquitination pathway. Alternate splicing results in multiple transcript variants.

Specifications

Antigen UBOX5
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ubox5
Gene Accession No. O94941
Gene Alias 1500010O06Rik; BOS_13392; C330018L13Rik; FAST kinase domain-containing protein 5; FAST kinase domains 5; FASTKD5; hUIP5; KIAA0860; LOC100344053; RGD1305440; RING finger protein 37; RING-type E3 ubiquitin transferase RNF37; RNF37; U box domain containing 5; UBCE7IP5; UbcM4-interacting protein 5; Ube7ip5; ubiquitin conjugating enzyme 7 interacting protein 5; Ubiquitin-conjugating enzyme 7-interacting protein 5; U-box domain containing 5; U-box domain-containing protein 5; UBOX5; UIP5
Gene Symbols Ubox5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human UBOX5/RNF37.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22888
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.