Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBP 301, Tocris Bioscience™

Catalog No. 176650
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Kainate antagonist

Potent kainate receptor antagonist (apparent Kd = 5.94 μM). Displays ∽ 30-fold selectivity over AMPA receptors.

Chemical Identifiers

CAS 569371-10-4
Molecular Formula C15H14IN3O6
Molecular Weight (g/mol) 459.196
InChI Key JHSYCOCOIYSZGI-NSHDSACASA-N
Synonym as-alpha-amino-3-4-carboxyphenyl methyl-3,4-dihydro-5-iodo-2,4-dioxo-1 2h-pyrimidinepropanoic acid, tocris-1766, s-1-2-amino-2-carboxyethyl-3-4-carboxybenzyl-5-iodopyrimidine-2,4-dione, ?s-?-amino-3-4-carboxyphenyl methyl-3,4-dihydro-5-iodo-2,4-dioxo-1 2h-pyrimidinepropanoic acid, 4-3-2s-2-amino-3-hydroxy-3-oxopropyl-5-iodo-2,6-dioxopyrimidin-1-yl methyl benzoic acid
PubChem CID 6604913
IUPAC Name 4-[[3-[(2S)-2-amino-2-carboxyethyl]-5-iodo-2,6-dioxopyrimidin-1-yl]methyl]benzoic acid
SMILES C1=CC(=CC=C1CN2C(=O)C(=CN(C2=O)CC(C(=O)O)N)I)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 459.2
Percent Purity >98%
Chemical Name or Material UBP 301
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.