Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UBP 302
GREENER_CHOICE

Catalog No. 207910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-791-0 10 mg
20-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-791-0 Supplier Tocris Bioscience™ Supplier No. 2079/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective kainate antagonist; active enantiomer of UBP 296 (Cat. No. 2078)

Potent and selective GluK1 (formally GluR5)-subunit containing kainate receptor antagonist (apparent KD = 402 nM); active enantiomer of UBP 296 (Cat. No. 2078). Displays ∽ 260-fold selectivity over AMPA receptors, ∽ 90-fold selectivity over recombinant human GluK2 (formally GluR6) and GluK5 (formally KA2)-containing kainate receptors and has little or no action at NMDA or group I mGlu receptors. Selectively blocks kainate receptor-mediated LTP induction in rat hippocampal mossy fibers. Also protects against soman-induced seizures and neuronal degeneration for up to 30 days when administered one hour after soman exposure.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 745055-91-8
Molecular Formula C15H15N3O6

Specifications

Quantity 10 mg
Formula Weight 333.3
Purity >98%
Molecular Weight (g/mol) 333.3
Chemical Name or Material UBP 302
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.