Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UBP 310
GREENER_CHOICE

Catalog No. 362110R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-211-0R 10 mg
36-215-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-211-0R Supplier Tocris Bioscience™ Supplier No. 3621/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

GluK1-selective kainate antagonist

UBP 310 is a GluK1 (formerly GLUK5) kainate receptor antagonist (IC50 = 130 nM); also blocks recombinant homomeric GluK3 (forrmerly GLUK7) receptors. Displays 12,700-fold selectivity for GluK1 (formerly GLUK5) over GluK2 (formerly GLUK6). Exhibits no activity at mGlu group I or NMDA receptors at concentrations of up to 10 μM. Apparent KD value is 18 ± 4 nM for depression of kainate responses on the dorsal root.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Specifications

Quantity 10 mg
Product Type UBP 310
Purity >98%
Molecular Weight (g/mol) 353.35
Inhibitors UBP 310

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.