Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBP1 Antibody (OTI9G7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274761H
Description
UBP1 Monoclonal specifically detects UBP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
UBP1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
UBP1 | |
Recombinant protein fragment corresponding to amino acids 47-269 of human UBP1 (NP_055332) produced in E.coli. | |
0.1 mL | |
7342.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI9G7 | |
Immunohistochemistry | |
DKFZp686L1745, LBP1, LBP-1a, LBP1A, LBP1B, LBP-1B, Transcription factor LBP-1, upstream binding protein 1 (LBP-1a), upstream-binding protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction