Learn More
UBP1 Mouse anti-Human, Clone: OTI8F1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808853

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
UBP1 functions as a transcriptional activator in a promoter context-dependent manner. Modulates the placental expression of CYP11A1. Involved in regulation of the alpha-globin gene in erythroid cells. Activation of the alpha-globin promoter in erythroid cells is via synergistic interaction with TFCP2. Involved in regulation of the alpha-globin gene in erythroid cells. Binds strongly to sequences around the HIV-1 initiation site and weakly over the TATA-box. Represses HIV-1 transcription by inhibiting the binding of TFIID to the TATA-box.Specifications
UBP1 | |
Monoclonal | |
Unconjugated | |
UBP1 | |
LBP-1B, LBP-1a, LBP1A, LBP1B | |
Mouse | |
Affinity Chromatography | |
RUO | |
7342 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8F1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NZI7 | |
UBP1 | |
Human recombit protein fragment corresponding to amino acids 47-269 of human UBP1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.