Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBP1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142532
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000. Preliminary analysis showed no signal in Western Blot.
UBP1 functions as a transcriptional activator in a promoter context-dependent manner. Modulates the placental expression of CYP11A1. Involved in regulation of the alpha-globin gene in erythroid cells. Activation of the alpha-globin promoter in erythroid cells is via synergistic interaction with TFCP2. Involved in regulation of the alpha-globin gene in erythroid cells. Binds strongly to sequences around the HIV-1 initiation site and weakly over the TATA-box. Represses HIV-1 transcription by inhibiting the binding of TFIID to the TATA-box.
Specifications
| UBP1 | |
| Polyclonal | |
| Unconjugated | |
| UBP1 | |
| Cp2b; LBP1; LBP1A; LBP-1a; LBP1B; LBP-1b; NF2d9; Nuclear factor 2d9; RGD1564700; transcription factor LBP-1; Ubp1; upstream binding protein 1; upstream binding protein 1 (LBP-1a); upstream-binding protein 1 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 7342 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9NZI7 | |
| UBP1 | |
| Peptide with sequence C-PNHQGDGASQTSGE (aa 349-362). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction