Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBP43Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501991769
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1769 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1769 Supplier Rockland Immunochemicals Supplier No. 600401476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against human UBP43 protein. A BLAST analysis was used to suggest reactivity with this protein from human, chimpanzee and orangutan based on 100% homology for the immunogen sequence. The core 7 amino acid sequence QPRERPR of the 10 amino acid immunogen sequence may be reactive with UBP43 from mouse and dog. This core sequence also shows partial homology with proteins other than UBP43. Cross reactivity with UBP43 homologues from other sources has not been determined.

USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.

Specifications

Antigen UBP43
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene USP18
Gene Accession No. Q9UMW8
Gene Alias 1110058H21Rik; 43 kDa ISG15-specific protease; AW047653; BOS_5660; bubp43; hUBP43; ISG15-specific-processing protease; ISG43; LOC100355501; ubiquitin specific peptidase 18; ubiquitin specific protease 15; ubiquitin specific protease 18; ubiquitin specific protease 18-like; Ubl carboxyl-terminal hydrolase 18; Ubl thioesterase 18; ubl thiolesterase 18; Ubp15; Ubp43; USP18
Gene Symbols USP18
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11274
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.