Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBQLN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110543 Supplier Invitrogen™ Supplier No. PA5110543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WTSTHGGSGS RQGRQDGDQD APDIRNRFPN FLGIIRLYDY LQQLHENPQS LGTYLQGTAS ALSQSQEPPP SVNRVPPSSP SSQEPGSGQP LPEESVAIKG RSSCPAFLRY PTENSTGQGG DQDGAGKSST GHSTNLPDLV SGLGDSANRV PFAPLSFSPT AAIPGIPEPP WLPSPAYPRS LRPDGMNPAP QLQDEIQPQL PLLMHLQAAM ANPRALQALR QIEQGLQVLA TEAPRLLLWF MPCLAGTGSV AGGIESREDP LMSEDPLPNP PPEVFPALDS AELGFLSPPF LHMLQDLVST NPQQLQPEAH FQVQL.

This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This gene is specifically expressed in the testis, and proposed to regulate cell-cycle progression during spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBQLN3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene UBQLN3
Gene Accession No. Q8C5U9, Q9H347
Gene Alias 4933400K24Rik; testicular tissue protein Li 220; TUP-1; ubiquilin 3; ubiquilin-3; Ubqln3
Gene Symbols UBQLN3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 306-620 of human UBQLN3 (NP_059509.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244178, 308905, 50613
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.