Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | UBR1 |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
UBR1 Polyclonal specifically detects UBR1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
UBR1 | |
Polyclonal | |
Rabbit | |
Human | |
E3 ubiquitin-protein ligase UBR1, E3a ligase, EC 6.3.2.-, JBS, MGC142065, MGC142067, N-recognin-1, ubiquitin ligase E3 alpha-I, ubiquitin protein ligase E3 component n-recognin 1, ubiquitin-protein ligase E3-alpha, Ubiquitin-protein ligase E3-alpha-1, Ubiquitin-protein ligase E3-alpha-I | |
UBR1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
197131 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:APEEEVTFDFYHKASRLGSSAMNIQMLLEKLKGIPQLEGQKDMITWILQMFDTVKRLREKSCLIVATTSGSESIKNDE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title