Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBR2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | UBR2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
UBR2 Polyclonal specifically detects UBR2 in Human samples. It is validated for Western Blot.Specifications
| UBR2 | |
| Polyclonal | |
| Rabbit | |
| Q8IWV8 | |
| 23304 | |
| Synthetic peptides corresponding to UBR2(ubiquitin protein ligase E3 component n-recognin 2) The peptide sequence was selected from the C terminal of UBR2. Peptide sequence QGLRRGNPLHLCKERFKKIQKLWHQHSVTEEIGHAQEANQTLVGIDWQHL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| bA49A4.1, C6orf133MGC71112, chromosome 6 open reading frame 133, dJ242G1.1, dJ392M17.3, DKFZp686C08114, E3 ubiquitin-protein ligase UBR2, EC 6.3.2.-, KIAA0349FLJ36357, N-recognin-2, RP3-392M17.3, ubiquitin protein ligase E3 component n-recognin 2, Ubiquitin-protein ligase E3-alpha-2, Ubiquitin-protein ligase E3-alpha-II | |
| UBR2 | |
| IgG | |
| 200 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title