Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537161
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537161 Supplier Invitrogen™ Supplier No. PA537161
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 200 kDa. Purity is >95% by SDS-PAGE.

The mammalian NUMB homolog plays a role in the determination of cell fate during development and binds with a variety of proteins, including Eps15, LNX1 and Notch 1. NumbL (NUMB-like protein), also known as Numb-R, NBL, CAG3A, CTG3a, NUMBLIKE or TNRC23, is a 609 amino acid cytoplasmic protein that, like NUMB, is thought to play a role in cell fate. Expressed at high levels in developing brain tissue, NumbL contains one PID (phosphotyrosine interaction domain) and plays an important role in neuronal differentiation, possibly associating with Eps15 and Notch 1. In mice, deletion of the NumbL gene is associated with early embryonic death, suggesting an essential role for NumbL in early development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBR2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene UBR2
Gene Accession No. Q6WKZ8, Q8IWV8
Gene Alias 9930021A08Rik; AI462103; AW540746; bA49A4.1; C6orf133; dJ242G1.1; dJ392M17.3; E130209G04Rik; E3 ubiquitin-protein ligase UBR2; Kiaa0349; mKIAA0349; N-recognin-2; RING-type E3 ubiquitin transferase UBR2; si:ch211-239i4.2; ubiquitin ligase E3 alpha-II; ubiquitin protein ligase E3 component n-recognin 2; ubiquitin-protein ligase E3-alpha-2; Ubiquitin-protein ligase E3-alpha-II; Ubr2
Gene Symbols UBR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human UBR2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224826, 23304, 363188
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.