Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542309
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542309 Supplier Invitrogen™ Supplier No. PA542309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GLKLLSWLGS IIGYSDGLRR ILCQVGLQEG PDGENSSLVD RLMLSDSKLW Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 91%.

The mammalian NUMB homolog plays a role in the determination of cell fate during development and binds with a variety of proteins, including Eps15, LNX1 and Notch 1. NumbL (NUMB-like protein), also known as Numb-R, NBL, CAG3A, CTG3a, NUMBLIKE or TNRC23, is a 609 amino acid cytoplasmic protein that, like NUMB, is thought to play a role in cell fate. Expressed at high levels in developing brain tissue, NumbL contains one PID (phosphotyrosine interaction domain) and plays an important role in neuronal differentiation, possibly associating with Eps15 and Notch 1. In mice, deletion of the NumbL gene is associated with early embryonic death, suggesting an essential role for NumbL in early development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBR2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UBR2
Gene Accession No. Q8IWV8
Gene Alias 9930021A08Rik; AI462103; AW540746; bA49A4.1; C6orf133; dJ242G1.1; dJ392M17.3; E130209G04Rik; E3 ubiquitin-protein ligase UBR2; Kiaa0349; mKIAA0349; N-recognin-2; RING-type E3 ubiquitin transferase UBR2; si:ch211-239i4.2; ubiquitin ligase E3 alpha-II; ubiquitin protein ligase E3 component n-recognin 2; ubiquitin-protein ligase E3-alpha-2; Ubiquitin-protein ligase E3-alpha-II; Ubr2
Gene Symbols UBR2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human UBR2 (aa 314-363).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23304
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.