Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBR2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 188531AP150UL
Description
The mammalian NUMB homolog plays a role in the determination of cell fate during development and binds with a variety of proteins, including Eps15, LNX1 and Notch 1. NumbL (NUMB-like protein), also known as Numb-R, NBL, CAG3A, CTG3a, NUMBLIKE or TNRC23, is a 609 amino acid cytoplasmic protein that, like NUMB, is thought to play a role in cell fate. Expressed at high levels in developing brain tissue, NumbL contains one PID (phosphotyrosine interaction domain) and plays an important role in neuronal differentiation, possibly associating with Eps15 and Notch 1. In mice, deletion of the NumbL gene is associated with early embryonic death, suggesting an essential role for NumbL in early development.Specifications
| UBR2 | |
| Polyclonal | |
| Unconjugated | |
| UBR2 | |
| C6orf133, KIAA0349, N recognin 2, RP3 392M17.3, UBR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 224826, 23304 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6WKZ8, Q8IWV8 | |
| Ubr2 | |
| UBR2 Fusion Protein Ag13446 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction