Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557255
Description
Immunogen sequence: DRSAWKHAGA LKKSTCDAEK SYEVLLSFVI SELFKGKLYH EEGTQECAMV NPIAWSPESM EKCLQDFCLP FLRITSLLQH HLFGEDLPSC QE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Specifications
UBR3 | |
Polyclonal | |
Unconjugated | |
UBR3 | |
E3 ubiquitin-protein ligase UBR3; KIAA2024; N-recognin-3; RING-type E3 ubiquitin transferase UBR3; ubiquitin protein ligase E3 component n-recognin 3 (putative); ubiquitin-protein ligase E3-alpha-3; Ubiquitin-protein ligase E3-alpha-III; UBR3; Zinc finger protein 650; ZNF650 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
130507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZT12 | |
UBR3 | |
Recombinant protein corresponding to Human UBR3. Recombinant protein control fragment (Product #RP-96127). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction