Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBR4 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128730F
Description
UBR4 Polyclonal antibody specifically detects UBR4 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
UBR4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
600 kDa retinoblastoma protein-associated factor, EC 6.3.2, EC 6.3.2.-, KIAA0462E3 ubiquitin-protein ligase UBR4, KIAA1307zinc finger, UBR1 type 1, N-recognin-4, p600, RBAF600FLJ41863, Retinoblastoma-associated factor of 600 kDa, ubiquitin protein ligase E3 component n-recognin 4, Zinc finger UBR1-type protein 1, ZUBR1retinoblastoma-associated factor 600-like protein | |
The immunogen recognized by this antibody maps to a region between residue 1350 and 1400 of human retinoblastoma-associated factor 600 using the numbering given in entry NP_065816.1 (GeneID 23352). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23352 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction