Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567591 Supplier Invitrogen™ Supplier No. PA567591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

E3 ubiquitin-protein ligase which is a component of the N-end rule pathway. Recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. Together with clathrin, forms meshwork structures involved in membrane morphogenesis and cytoskeletal organization. Regulates integrin-mediated signaling. May play a role in activation of FAK in response to cell-matrix interactions. Mediates ubiquitination of ACLY, leading to its subsequent degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBR4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UBR4
Gene Accession No. A2AN08, Q2TL32, Q5T4S7
Gene Alias 1810009A16Rik; 600 kDa retinoblastoma protein-associated factor; A930005E13Rik; D930005K06Rik; E3 ubiquitin-protein ligase UBR4; Gm1032; Gm1666; KIAA0462; KIAA1307; mKIAA0462; N28143; N-recognin-4; p600; Rbaf600; retinoblastoma-associated factor 600; retinoblastoma-associated factor 600-like protein; retinoblastoma-associated factor of 600 kDa; RING-type E3 ubiquitin transferase UBR4; ubiquitin protein ligase E3 component n-recognin 4; UBR4; Zinc finger UBR1-type protein 1; zinc finger, UBR1 type 1; ZUBR1
Gene Symbols UBR4
Host Species Rabbit
Immunogen Synthesized peptide derived from human UBR4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23352, 313658, 69116
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.