Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116101
Description
Antibody detects endogenous levels of total UBR4.
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway. Recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. Together with clathrin, forms meshwork structures involved in membrane morphogenesis and cytoskeletal organization. Regulates integrin-mediated signaling. May play a role in activation of FAK in response to cell-matrix interactions. Mediates ubiquitination of ACLY, leading to its subsequent degradation.
Specifications
| UBR4 | |
| Polyclonal | |
| Unconjugated | |
| UBR4 | |
| 1810009A16Rik; 600 kDa retinoblastoma protein-associated factor; A930005E13Rik; D930005K06Rik; E3 ubiquitin-protein ligase UBR4; Gm1032; Gm1666; KIAA0462; KIAA1307; mKIAA0462; N28143; N-recognin-4; p600; Rbaf600; retinoblastoma-associated factor 600; retinoblastoma-associated factor 600-like protein; retinoblastoma-associated factor of 600 kDa; RING-type E3 ubiquitin transferase UBR4; ubiquitin protein ligase E3 component n-recognin 4; UBR4; Zinc finger UBR1-type protein 1; zinc finger, UBR1 type 1; ZUBR1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23352, 313658, 69116 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| A2AN08, Q2TL32, Q5T4S7 | |
| UBR4 | |
| A synthesized peptide derived from human UBR4(Accession Q5T4S7), corresponding to amino acid residues P4006-A4056. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction