Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBR5/EDD Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171802AF700
Description
UBR5/EDD Polyclonal antibody specifically detects UBR5/EDD in Human, Mouse samples. It is validated for Western BlotSpecifications
UBR5/EDD | |
Polyclonal | |
Western Blot | |
DD5, E3 ubiquitin protein ligase, HECT domain containing, 1, E3 ubiquitin-protein ligase, HECT domain-containing 1, EC 6.3.2, EC 6.3.2.-, EDD1E3 ubiquitin-protein ligase UBR5, EDDE3 identified by differential display, hHYD, HYDFLJ11310, Hyperplastic discs protein homolog, KIAA0896MGC57263, progestin induced protein, Progestin-induced protein, ubiquitin protein ligase E3 component n-recognin 5, ubiquitin-protein ligase | |
The immunogen for this product maps to a region between residue 2749 and 2799 of human E3 Ubiquitin Protein Ligase, HECT domain using the numbering given in entry NP_056986.2 (GeneID 51366). | |
0.1 mL | |
DNA Repair | |
51366 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction