Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBR5/EDD Antibody (SR1501), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322217100UL
Description
UBR5/EDD Monoclonal antibody specifically detects UBR5/EDD in Human, Mouse samples. It is validated for Western BlotSpecifications
UBR5/EDD | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
SR1501 | |
Western Blot 1:500-1:2000 | |
DD5, E3 ubiquitin protein ligase, HECT domain containing, 1, E3 ubiquitin-protein ligase, HECT domain-containing 1, EC 6.3.2, EC 6.3.2.-, EDD1E3 ubiquitin-protein ligase UBR5, EDDE3 identified by differential display, hHYD, HYDFLJ11310, Hyperplastic discs protein homolog, KIAA0896MGC57263, progestin induced protein, Progestin-induced protein, ubiquitin protein ligase E3 component n-recognin 5, ubiquitin-protein ligase | |
A synthesized peptide derived from human UBR5/EDD (Uniprot #: O95071) | |
100 μg | |
DNA Repair | |
51366 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction