Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBTD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557075
Description
Immunogen sequence: YQLPIYCLSP PVNLLLEHTE EESLEPPEPP PSVRREFPLK VRLSTGKDVR LSASLPDTVG QLKRQLHAQE GIEPSWQRWF FSGKLLT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
The degradation of many proteins is carried out by the ubiquitin pathway in which proteins are targeted for degradation by covalent conjugation of the polypeptide ubiquitin. This gene encodes a protein that belongs to the ubiquitin family of proteins. The encoded protein is thought to regulate E2 ubiquitin conjugating enzymes belonging to the UBE2D family.
Specifications
| UBTD1 | |
| Polyclonal | |
| Unconjugated | |
| UBTD1 | |
| BC016129; FLJ11807; ubiquitin domain containing 1; ubiquitin domain-containing protein 1; Ubtd1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80019 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HAC8 | |
| UBTD1 | |
| Recombinant protein corresponding to Human UBTD1. Recombinant protein control fragment (Product #RP-97601). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction