Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBTD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557449 Supplier Invitrogen™ Supplier No. PA557449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLPVYCLAPP INMIEEKSDI ETLDIPEPPP NSGYECQLRL RLSTGKDLKL VVRSTD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

UBTD2 gene ontology (GO) annotation includes cytoplasm and cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBTD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBTD2
Gene Accession No. Q8WUN7
Gene Alias 4930571L24; 9630054F20Rik; AI645720; DCUBP; DC-UbP; Dendritic cell-derived ubiquitin-like protein; RGD1305506; SB72; ubiquitin domain containing 2; ubiquitin domain-containing protein 2; Ubiquitin-like protein SB72; Ubtd2
Gene Symbols UBTD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBTD2. Recombinant protein control fragment (Product #RP-102686).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.