Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubtfl1 Polyclonal Antibody
Ubtfl1 Polyclonal Antibody
Supplier: Thermo Scientific PA549394
Description
Ubtfl1 Polyclonal Antibody for Western Blot

Specifications
Ubtfl1 | |
Polyclonal | |
Unconjugated | |
Ubtfl1 | |
B020006M18Rik; C11orf27; HMG-box preimplantation embryo-specific protein; HMG-box protein, preimplantation-embryo-specific; HMGPI; putative upstream-binding factor 1-like protein 1; Ubtfl1; upstream binding transcription factor, RNA polymerase I-like 1; upstream-binding factor 1-like protein 1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
546118 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q3USZ2 | |
Ubtfl1 | |
KLH conjugated synthetic peptide between 186-220 amino acids from the Central region of mouse Ubtfl1. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction