Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBXD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557611 Supplier Invitrogen™ Supplier No. PA557611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERSTVARIQF RLPDGSSFTN QFPSDAPLEE ARQFAAQTVG NTYGNFSLAT MFPRREFTKE DYKKKLLDLE LAPSASVVLL PAGRPTASIV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

MAFA is a transcription factor that binds RIPE3b, a conserved enhancer element that regulates pancreatic beta cell-specific expression of the insulin gene. MAFA cooperates synergistically with NEUROD1 and PDX1. Phosphorylation by GSK3 increases its transcriptional activity and is required for its ocogenic activity. Serves as an ocogene or tumor supressor depending on the cell context.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBXD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBXN4
Gene Accession No. Q5HZY0, Q8VCH8, Q92575
Gene Alias 1300013G12Rik; erasin; hypothetical protein LOC536181; KIAA0242; mKIAA0242; UBX domain containing 2; UBX domain protein 4; UBX domain-containing 1; UBX domain-containing protein 2; UBX domain-containing protein 4; UBXD2; UBXDC1; ubxn4; zgc:73362
Gene Symbols UBXN4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBXD2. Recombinant protein control fragment (Product #RP-96121).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23190, 304766, 67812
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.