Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBXD5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | UBXD5 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
UBXD5 Polyclonal specifically detects UBXD5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
UBXD5 | |
Polyclonal | |
Rabbit | |
Human | |
91544 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RLYPNGVPFKVSDLRNQVYLEDGLDPFPGEGRVVGRQLMHKALDRVEEHPGSRMTAEKFLNRLPKFVIRQGEVIDIRGPIRDTLQNCCPLPARIQEIVVET | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
COA-1, Colorectal tumor-associated antigen COA-1, colorectal tumor-associated antigen-1, SOCDKFZp686F04228, SOCI, socius, UBX domain containing 5, UBX domain protein 11, UBX domain-containing protein 11, UBX domain-containing protein 5, UBXD5 | |
UBXN11 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title