Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565095 Supplier Thermo Scientific Supplier No. PA565095
Add to Cart
Add to Cart

UBXD5 Polyclonal Antibody

UBXD5 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen UBXD5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBXN11
Gene Accession No. Q5T124
Gene Alias 4930506L07Rik; COA-1; colorectal tumor-associated antigen COA-1; colorectal tumor-associated antigen-1; D4Bwg1540e; PP2243; Soc; SOCI; Socius; UBX domain containing 5; UBX domain protein 11; UBX domain-containing protein 11; UBX domain-containing protein 5; UBXD5; Ubxn11
Gene Symbols UBXN11
Host Species Rabbit
Immunogen Recombinant Human UBXD5. Recombinant protein control fragment (Product #RP-104629).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91544
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.