Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBXD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104520
Description
Antibody detects endogenous levels of total UBXD5.
This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3' coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
UBXD5 | |
Polyclonal | |
Unconjugated | |
UBXN11 | |
4930506L07Rik; COA-1; colorectal tumor-associated antigen COA-1; colorectal tumor-associated antigen-1; D4Bwg1540e; PP2243; Soc; SOCI; Socius; UBX domain containing 5; UBX domain protein 11; UBX domain-containing protein 11; UBX domain-containing protein 5; UBXD5; Ubxn11 | |
Rabbit | |
Affinity chromatography | |
RUO | |
192207, 67586, 91544 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5T124, Q8R512, Q9D572 | |
UBXN11 | |
A synthesized peptide derived from human UBXN11(Accession Q5T124), corresponding to amino acid residues R138-D188. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction