Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBXN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552967 Supplier Invitrogen™ Supplier No. PA552967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAELTALESL IEMGFPRGRA EKALALTGNQ GIEAAMDWLM EHEDDPDVDE PLETPLGHIL GREPTSSEQG GLEGSGSAAG EGKPALSEEE RQEQTKRMLE LVAQKQRERE EREEREALER ERQRRRQGQE LSAARQRLQE DEMRRAAEER Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

Ubiquitin-binding protein that plays a role in the modulation of innate immune response. Blocks both the RIG-I-like receptors (RLR) and NF-kappa-B pathways. Following viral infection, UBXN1 is induced and recruited to the RLR component MAVS. In turn, interferes with MAVS oligomerization, and disrupts the MAVS/TRAF3/TRAF6 signalosome. This function probably serves as a brake to prevent excessive RLR signaling (PubMed:23545497). Interferes with the TNFalpha-triggered NF-kappa-B pathway by interacting with cellular inhibitors of apoptosis proteins (cIAPs) and thereby inhibiting their recruitment to TNFR1 (PubMed:25681446). Prevents also the activation of NF-kappa-B by associating with CUL1 and thus inhibiting NF-kappa-B inhibitor alpha/NFKBIA degradation that remains bound to NF-kappa-B (PubMed:28152074). Interacts with the BRCA1-BARD1 heterodimer and regulates its activity. Specifically binds 'Lys-6'-linked polyubiquitin chains. Interaction with autoubiquitinated BRCA1 leads to the inhibition of the E3 ubiquitin-protein ligase activity of the BRCA1-BARD1 heterodimer (PubMed:20351172). Component of a complex required to couple deglycosylation and proteasome-mediated degradation of misfolded proteins in the endoplasmic reticulum that are retrotranslocated in the cytosol. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBXN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBXN1
Gene Accession No. Q04323, Q499N6, Q922Y1
Gene Alias 2B28; 4930455J02Rik; D19Ertd721e; mY33K; Protein 2B28; RGD1309471; SAKS1; SAPK substrate protein 1; T25529; UBA/UBX 33.3 kDa protein; UBX domain protein 1; UBX domain-containing protein 1; UBXD10; UBXN1
Gene Symbols UBXN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBXN1. Recombinant protein control fragment (Product #RP-88780).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225896, 293719, 51035
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.