Learn More
UBXN1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303415AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Ubiquitin-binding protein that interacts with the BRCA1-BARD1 heterodimer, and regulates its activity. Specifically binds ′Lys-6′-linked polyubiquitin chains. Interaction with autoubiquitinated BRCA1, leads to inhibit the E3 ubiquitin-protein ligase activity of the BRCA1-BARD1 heterodimer. Component of a complex required to couple deglycosylation and proteasome-mediated degradation of misfolded proteins in the endoplasmic reticulun that are retrotranslocated in the cytosol.Specifications
UBXN1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q04323 | |
UBXN1 | |
Between 50 and 100 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
UBXN1 | |
2B28, SAKS1, UBXD10, SAPK substrate protein 1, UBA/UBX 33.3 kDa protein, UBX domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51035 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.