Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBXN10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$377.03 - $666.47
Specifications
| Antigen | UBXN10 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
UBXN10 Polyclonal specifically detects UBXN10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| UBXN10 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ25429, UBX domain containing 3, UBX domain protein 10, UBX domain-containing protein 10, UBX domain-containing protein 3, UBXD3 | |
| UBXN10 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 127733 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RFVRHFRPTDDLQTIVAVAEQKNKTSYRHCSIETMEVPRRRFSDLTKSLQECRIPHKSVLGISLEDGEGW | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title