Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBXN10 Antibody (OTI1H5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274762H
Description
UBXN10 Monoclonal specifically detects UBXN10 in Human samples. It is validated for Western Blot.Specifications
UBXN10 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
UBXN10 | |
Full length recombinant protein of human UBXN10 (NP_689589) produced in HEK293T cells. | |
0.1 mL | |
127733.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1H5 | |
Western Blot | |
FLJ25429, UBX domain containing 3, UBX domain protein 10, UBX domain-containing protein 10, UBX domain-containing protein 3, UBXD3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction