Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBXN10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555897
Description
Immunogen sequence: QLSSIRALYQ DETGTMKTSE EDSRARACAV ERKFIVRTKK QGSSRAGNLE EPSDQEPRLL LAVRSPTGQ Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 69%.
UBXN10 (VCP/p97) binding protein required for ciliogenesis. Acts as a tethering factor that facilitates recruitment of VCP/p97 to the intraflagellar transport complex B (IFT-B) in cilia. UBX domain-containing proteins act as tethering factors for VCP/p97 and may specify substrate specificity of VCP/p97.
Specifications
UBXN10 | |
Polyclonal | |
Unconjugated | |
Ubxn10 | |
5730509E04Rik; A830047D02; UBX domain containing 3; UBX domain protein 10; UBX domain-containing protein 10; UBX domain-containing protein 3; UBXD3; UBXN10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LJ8 | |
Ubxn10 | |
Recombinant protein corresponding to Human UBXN10. Recombinant protein control fragment (Product #RP-94373). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction