Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBXN2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564224
Description
Immunogen sequence: MKDVDNLKSI KEEWVCETGS DNQPLGNNQQ SNCEYFVDSL FEEAQKVSSK CVSPAEQKKQ VDVN Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 77%.
UBXN2A gene ontology annotations related to this gene include autophagosome assembly; cellular response to leukemia inhibitory factor; Golgi organization; membrane fusion; nuclear envelope reassembly; proteasome-mediated ubiquitin-dependent protein catabolic process; regulation of gene expression; regulation of protein catabolic process; regulation of protein ubiquitination.
Specifications
UBXN2A | |
Polyclonal | |
Unconjugated | |
UBXN2A | |
6330407P03Rik; UBX domain containing 4; UBX domain protein 2A; UBX domain-containing protein 2A; UBX domain-containing protein 4; Ubxd4; Ubxn2a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
165324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P68543 | |
UBXN2A | |
Recombinant protein corresponding to Human UBXN2A. Recombinant protein control fragment (Product #RP-103870). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction