Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBXN2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569953
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
UBXN2A gene ontology annotations related to this gene include autophagosome assembly; cellular response to leukemia inhibitory factor; Golgi organization; membrane fusion; nuclear envelope reassembly; proteasome-mediated ubiquitin-dependent protein catabolic process; regulation of gene expression; regulation of protein catabolic process; regulation of protein ubiquitination.
Specifications
UBXN2A | |
Polyclonal | |
Unconjugated | |
UBXN2A | |
6330407P03Rik; UBX domain containing 4; UBX domain protein 2A; UBX domain-containing protein 2A; UBX domain-containing protein 4; Ubxd4; Ubxn2a | |
Rabbit | |
Affinity chromatography | |
RUO | |
685859 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
UBXN2A | |
synthetic peptide corresponding to a region of rat UBXN2A. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction