Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBXN2B Antibody (OTI1F7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200543
Description
UBXN2B Monoclonal specifically detects UBXN2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
UBXN2B | |
Monoclonal | |
0.41 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
UBXN2B | |
Full length human recombinant protein of human UBXN2B (NP_001071087) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1F7 | |
Unconjugated | |
UBX domain protein 2B | |
Mouse | |
36.9 kDa | |
0.1 mL | |
Signal Transduction | |
137886 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction