Learn More
Invitrogen™ UBXN2B Monoclonal Antibody (OTI1G3)

Description
UBXN2B Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | UBXN2B |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI1G3 |
| Concentration | 0.39 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | UBXN2B |
| Gene Accession No. | Q14CS0 |
| Gene Alias | NSFL1 cofactor p37; p37; p97 cofactor p37; UBX domain protein 2B; UBX domain-containing protein 2B; UBXN2B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.