Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBXN2B Monoclonal Antibody (OTI2E8), TrueMAB™, OriGene
SDP

Catalog No. 502541516
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1516 30 μL
50-167-3535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1516 Supplier OriGene Supplier No. TA502483S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBXN2B Monoclonal antibody specifically detects UBXN2B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen UBXN2B
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2E8
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ndufb9
Gene Accession No. Q9Y6M9
Gene Alias NSFL1 cofactor p37; p37; p97 cofactor p37; UBX domain protein 2B; UBX domain-containing protein 2B; UBXN2B
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 3-179 of human NDUFB9 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4715
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.