Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBXN2B Antibody (OTI1F7), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274769PCP
Description
UBXN2B Monoclonal specifically detects UBXN2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
UBXN2B | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
UBXN2B | |
Full length recombinant protein of human UBXN2B (NP_001071087) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI1F7 | |
Flow Cytometry | |
UBX domain protein 2B | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
137886.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction