Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBXN6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559511 Supplier Invitrogen™ Supplier No. PA559511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDTIRNQVRK ELQAEATVSG SPEAPGTNVV SEPREEGSAH LAVPGVYFTC PLTGATLRKD QRDACIKEAI LLHFSTDPV Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.

May negatively regulate the ATPase activity of VCP, an ATP-driven segregase that associates with different cofactors to control a wide variety of cellular processes (PubMed:26475856). As a cofactor of VCP, it may play a role in the transport of CAV1 to lysosomes for degradation (PubMed:21822278, PubMed:23335559). It may also play a role in endoplasmic reticulum-associated degradation (ERAD) of misfolded proteins (PubMed:19275885). Together with VCP and other cofactors, it may play a role in macroautophagy, regulating for instance the clearance of damaged lysosomes (PubMed:27753622). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBXN6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBXN6
Gene Accession No. Q9BZV1
Gene Alias 1200008L11Rik; 2210415J11Rik; AU040909; CTB-50L17.16; UBX domain containing 1; UBX domain protein 6; UBX domain-containing 1; UBX domain-containing 2; UBX domain-containing protein 1; UBX domain-containing protein 6; UBXD1; Ubxdc2; Ubxn6
Gene Symbols UBXN6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBXN6. Recombinant protein control fragment (Product #RP-99154).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80700
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.