Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBXN7 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222223DL594
Description
UBXN7 Polyclonal antibody specifically detects UBXN7 in Human, Mouse samples. It is validated for Western BlotSpecifications
UBXN7 | |
Polyclonal | |
Western Blot | |
KIAA0794, UBX domain containing 7, UBX domain protein 7, UBX domain-containing protein 7, UBXD7 | |
The immunogen this antibody was made to, maps to a region between residue 439 and 489 of human UBX domain protein 7 using the numbering given in entry NP_056377.1 (GeneID 26043). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
26043 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction