Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBXN8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582595 Supplier Invitrogen™ Supplier No. PA582595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDEGTSQTSF ETSNREAAKS QNLPKPLTEF PSPAEQPTCK EIPDLPEEPS QTAEEVVTVA LRCPSGNVLR RRFLKSYSSQ VLFDWMTRIG YHISLYSLST SFPRRPLAVE GGQSLEDIGI TVDTVLILEE KEQTN Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 73%.

Involved in endoplasmic reticulum-associated degradation (ERAD) for misfolded lumenal proteins, possibly by tethering VCP to the endoplasmic reticulum membrane. May play a role in reproduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBXN8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBXN8
Gene Accession No. O00124
Gene Alias D8S2298E; REP8; Rep-8 protein; Reproduction 8 protein; Reproduction/chromosome 8; UBX domain protein 8; UBX domain-containing protein 6; UBX domain-containing protein 8; UBXD6; UBXN8
Gene Symbols UBXN8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBXN8. Recombinant protein control fragment (Product #RP-108151).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7993
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.