Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCH-L1/PGP9.5 Antibody (31A3) [Janelia Fluor™ 525], Novus Biologicals™
SDP

Catalog No. NB801156 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB801156 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB801156 Supplier Novus Biologicals Supplier No. NBP233130JF525
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UCH-L1/PGP9.5 Monoclonal antibody specifically detects UCH-L1/PGP9.5 in Bovine,Canine,Human,Mouse,Porcine,Rat,Zebrafish samples. It is validated for Flow Cytometry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Simple Western,Immunocytochemistry/ Immunofluorescence

Specifications

Antigen UCH-L1/PGP9.5
Applications Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Simple Western, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 31A3
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
Host Species Mouse
Immunogen UCH-L1/PGP9.5 protein from brain (Uniprot: P09936)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 7345
Target Species Bovine, Canine, Human, Mouse, Pig, Rat, Zebrafish
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.