Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCH-L1/PGP9.5 Antibody (BH7), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11058869AF532
Description
UCH-L1/PGP9.5 Monoclonal antibody specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry Free-FloatingSpecifications
UCH-L1/PGP9.5 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Equine | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
BH7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating | |
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
Recombinant full length human UCHL1 purified from E. coli. [Swiss-Prot# P09936] | |
0.1 mL | |
Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
7345 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction