Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCH-L1/PGP9.5 Antibody (UCHL1/7076R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32028020UG
Description
UCH-L1/PGP9.5 Monoclonal antibody specifically detects UCH-L1/PGP9.5 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
UCH-L1/PGP9.5 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 2-4 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
Recombinant full-length human UCH-L1/PGP9.5 protein | |
20 μg | |
Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
7345 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Western Blot, Immunohistochemistry (Paraffin) | |
UCHL1/7076R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction