Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCH-L1/PGP9.5 Antibody (UCHL1/841), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | UCH-L1/PGP9.5 |
---|---|
Clone | UCHL1/841 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
Applications | Western Blot, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24479600
![]() |
Novus Biologicals
NBP2447960.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24479601
![]() |
Novus Biologicals
NBP2447960.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24479602
![]() |
Novus Biologicals
NBP2447960.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Western Blot, Immunofluorescence
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human samples. It is validated for Western Blot.Specifications
UCH-L1/PGP9.5 | |
0.2 mg/mL | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Mouse | |
Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
7345 | |
Recombinant human UCHL1 protein | |
Primary | |
Store at 4C. |
UCHL1/841 | |
Western Blot 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
UCHL1 | |
IgG2a κ | |
Protein A or G purified | |
This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title