Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCH-L3 Antibody (033), HRP

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290678H
Description
UCH-L3 Monoclonal specifically detects UCH-L3 in Mouse samples. It is validated for ELISA.Specifications
UCH-L3 | |
Monoclonal | |
HRP | |
EC 3.4.19.12, ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, ubiquitin thiolesterase, UCH-L3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse UCH-L3 (Q9JKB1; Glu2-Ala230). | |
0.1 mL | |
Primary | |
Has cross-reactivity in ELISA with: Human UCHL3 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
33 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
7347 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction