Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCH-L3 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298421IR
Description
UCH-L3 Polyclonal specifically detects UCH-L3 in Rat samples. It is validated for Western Blot, ELISA.Specifications
UCH-L3 | |
Polyclonal | |
Western Blot, ELISA | |
EC 3.4.19.12, ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, ubiquitin thiolesterase, UCH-L3 | |
Produced in rabbits immunized with purified, recombinant UCH-L3 (Q91Y78; Glu 2-Ala 230). | |
0.1 mL | |
Primary | |
Rat | |
Purified |
Western Blot, ELISA | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
7347 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction