Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCH-L3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563877
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3877 Supplier Bethyl Laboratories, Inc Supplier No. A302947A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine and Pig

The two ubiquitin C-terminal hydrolase (UCH) enzymes, UCHL1 and UCHL3, deubiquitinate ubiquitin-protein conjugates and control the cellular balance of ubiquitin. UCHL1 and UCHL3 are both small proteins of ∽220 amino acids that share more than 40% amino acid sequence identity. UCHL3 is universally expressed in all tissues, while UCHL1 is expressed exclusively in neuronal tissue, testis and ovary. The activity of UCHL3 is more than 200 fold higher than UCHL1 when a fluorogenic ubiquitin substrate is used. UCHL1 associates with monoubiquitin and UCHL3 binds to Nedd8, ubiquitin-like protein. UCHL1 and UCHL3 play a role in the regulation of neuronal development and spermatogenesis. UCHL1 is involved in the pathogenesis of Parkinson′s disease (PD) and Alzheimer′s disease (AD). Down-regulation and extensive oxidative modification of UCHL1 have been observed in the brains of AD patients as well as PD patients.

Specifications

Antigen UCH-L3
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene UCHL3
Gene Accession No. P15374
Gene Alias ubiquitin carboxyl-terminal hydrolase isozyme L3; ubiquitin thioesterase L3; ubiquitin thiolesterase
Gene Symbols UCHL3
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7347
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.